1-(4-(tert-Butyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140189-1G 140189-500MG 140189-5G 140189-25G
77464-840EA 415.09 USD
77464-840 77477-618 77487-734 77498-462
1-(4-(tert-Butyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-(tert-Butyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787359
CAS Number: 34235-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR