2,2-Dimethyl-1-(4-propoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140183-1G 140183-500MG 140183-5G 140183-25G
77464-836EA 415.09 USD
77464-836 77477-614 77487-730 77498-458
2,2-Dimethyl-1-(4-propoxyphenyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(4-propoxyphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17224006
CAS Number: 1343643-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR