1-(3-Isopropoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140179-1G 140179-500MG 140179-5G
77464-832EA 447.02 USD
77464-832 77477-610 77487-726
1-(3-Isopropoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Isopropoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD21328301
CAS Number: 1491992-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR