2,2-Dimethyl-1-(m-tolyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140173-1G 140173-500MG 140173-5G 140173-25G
77464-828EA 415.09 USD
77464-828 77477-606 77487-722 77498-454
2,2-Dimethyl-1-(m-tolyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(m-tolyl)propan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153733
CAS Number: 55678-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR