1-(2-Ethoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140161-1G 140161-500MG 140161-5G 140161-25G
77464-824EA 347.11 USD
77464-824 77477-602 77487-718 77498-450
1-(2-Ethoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Ethoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17223687
CAS Number: 1342247-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR