1-(3-Chloro-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139921-1G 139921-500MG 139921-5G 139921-25G
77464-816EA 415.09 USD
77464-816 77477-594 77487-710 77498-444
1-(3-Chloro-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(3-Chloro-4-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇ClO
MW: 212.7157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29501681
CAS Number: 1594567-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR