1-(2-Fluoro-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139919-1G 139919-500MG 139919-5G 139919-25G
77464-812EA 397.58 USD
77464-812 77477-590 77487-706 77498-440
1-(2-Fluoro-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(2-Fluoro-4-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.2611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34147531
CAS Number: 1601028-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR