1-(4-Fluoro-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139917-1G 139917-500MG 139917-5G 139917-25G
77464-808EA 415.09 USD
77464-808 77477-586 77487-702 77498-436
1-(4-Fluoro-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(4-Fluoro-2-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO
Storage Temperature: Ambient
MDL Number: MFCD17222676
CAS Number: 1344376-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR