2-Methyl-1-(4-propylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139904-1G 139904-500MG 139904-5G 139904-25G
77464-804EA 347.11 USD
77464-804 77477-582 77487-698 77498-432
2-Methyl-1-(4-propylphenyl)propan-1-ol ≥95%
2-Methyl-1-(4-propylphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787416
CAS Number: 1216265-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR