1-(3-Chloro-4-methylphenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139834-1G 139834-500MG 139834-5G 139834-25G
77464-792EA 415.09 USD
77464-792 77477-570 77487-686 77498-424
1-(3-Chloro-4-methylphenyl)-2-methylpropan-1-ol ≥97%
1-(3-Chloro-4-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅ClO
MW: 198.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34834260
CAS Number: 1602743-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR