1-(2-Isopropylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139800-1G 139800-500MG 139800-5G 139800-25G
77464-784EA 397.58 USD
77464-784 77477-562 77487-678 77498-422
1-(2-Isopropylphenyl)-2-methylpropan-1-ol ≥95%
1-(2-Isopropylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30070942
CAS Number: 39615-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR