1-(3,5-dimethylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139772-1G 139772-500MG 139772-5G 139772-25G
77464-780EA 415.09 USD
77464-780 77477-558 77487-674 77498-418
1-(3,5-dimethylphenyl)-2-methylpropan-1-ol ≥95%
1-(3,5-dimethylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12806387
CAS Number: 164394-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR