1-(4-Ethylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139744-1G 139744-500MG 139744-5G 139744-25G
77464-776EA 415.09 USD
77464-776 77477-554 77487-670 77498-414
1-(4-Ethylphenyl)-2-methylpropan-1-ol ≥95%
1-(4-Ethylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934820
CAS Number: 68857-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR