2-Methyl-1-(o-tolyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139739-1G 139739-500MG 139739-5G 139739-25G
77464-772EA 363.59 USD
77464-772 77477-550 77487-666 77498-410
2-Methyl-1-(o-tolyl)propan-1-ol ≥95%
2-Methyl-1-(o-tolyl)propan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₆O
MW: 164.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153670
CAS Number: 100994-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR