phenyl(3-propoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139693-1G 139693-500MG 139693-5G
77464-768EA 447.02 USD
77464-768 77477-546 77487-662
phenyl(3-propoxyphenyl)methanol ≥95%
phenyl(3-propoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD18037541
CAS Number: 1281898-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR