(2,5-Dimethoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139646-1G 139646-500MG 139646-5G 139646-25G
77464-760EA 415.09 USD
77464-760 77477-538 77487-654 77498-402
(2,5-Dimethoxyphenyl)(phenyl)methanol ≥95%
(2,5-Dimethoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD13474845
CAS Number: 98635-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR