2-Chloro-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

138536-1G 138536-500MG 138536-5G
77464-756EA 594.31 USD
77464-756 77477-536 77487-650
2-Chloro-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₇ClF₂O₃
MW: 248.611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045428
CAS Number: 1565025-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR