2-Chloro-1-(2,4-dichlorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

138456-1G 138456-500MG 138456-5G
77464-752EA 576.8 USD
77464-752 77477-532 77487-646
2-Chloro-1-(2,4-dichlorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,4-dichlorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₃Cl₃F₂O
MW: 259.465 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28105064
CAS Number: 121412-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR