1-(4-butylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

138443-1G 138443-500MG 138443-5G
77464-744EA 576.8 USD
77464-744 77477-524 77487-638
1-(4-butylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(4-butylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₂H₁₃ClF₂O
MW: 246.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920301
CAS Number: 1352226-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR