2-Chloro-2,2-difluoro-1-(4-isopropoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138432-1G 138432-500MG 138432-5G
77464-740EA 696.28 USD
77464-740 77477-520 77487-634
2-Chloro-2,2-difluoro-1-(4-isopropoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4-isopropoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁ClF₂O₂
MW: 248.654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045569
CAS Number: 1566459-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR