2-Chloro-2,2-difluoro-1-(p-tolyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138425-1G 138425-500MG 138425-5G
77464-736EA 576.8 USD
77464-736 77477-516 77487-630
2-Chloro-2,2-difluoro-1-(p-tolyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(p-tolyl)ethanone

Precaución: For research use only

Formula: C₉H₇ClF₂O
MW: 204.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13173017
CAS Number: 89264-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR