2-Chloro-2,2-difluoro-1-(4-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138421-1G 138421-500MG 138421-5G
77464-732EA 546.93 USD
77464-732 77477-512 77487-626
2-Chloro-2,2-difluoro-1-(4-propoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁ClF₂O₂
MW: 248.654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045567
CAS Number: 1564694-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR