2-Chloro-2,2-difluoro-1-(3-isopropoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138416-1G 138416-500MG 138416-5G
77464-728EA 576.8 USD
77464-728 77477-508 77487-622
2-Chloro-2,2-difluoro-1-(3-isopropoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-isopropoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26045479
CAS Number: 1566709-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR