2-Chloro-2,2-difluoro-1-(3-isopropylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138412-1G 138412-500MG 138412-5G
77464-724EA 594.31 USD
77464-724 77477-504 77487-618
2-Chloro-2,2-difluoro-1-(3-isopropylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-isopropylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁ClF₂O
MW: 232.654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103965
CAS Number: 1823316-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR