2-Chloro-2,2-difluoro-1-(3-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138409-1G 138409-500MG 138409-5G
77464-720EA 594.31 USD
77464-720 77477-500 77487-614
2-Chloro-2,2-difluoro-1-(3-methoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-methoxyphenyl)ethanone

Precaución: For research use only

Formula: C₉H₇ClF₂O₂
MW: 220.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20921142
CAS Number: 1352225-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR