2-Chloro-2,2-difluoro-1-(2-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138392-1G 138392-500MG 138392-5G
77464-716EA 576.8 USD
77464-716 77477-496 77487-610
2-Chloro-2,2-difluoro-1-(2-(trifluoromethyl)phenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-(trifluoromethyl)phenyl)ethanone

Precaución: For research use only

Formula: C₉H₄ClF₅O
MW: 258.572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045465
CAS Number: 1564717-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR