8-(4-(Methylthio)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

137752-1G 137752-500MG 137752-5G
77464-712EA 546.93 USD
77464-712 77477-492 77487-606
8-(4-(Methylthio)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(4-(Methylthio)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₂₀O₃S
MW: 280.382 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34833014
CAS Number: 1252615-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR