8-(2,6-Difluoropyridin-3-yl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

137486-1G 137486-500MG 137486-5G 137486-25G
77464-704EA 415.09 USD
77464-704 77477-484 77487-598 77498-382
8-(2,6-Difluoropyridin-3-yl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2,6-Difluoropyridin-3-yl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₃H₁₅F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD34832859
CAS Number: 2969464-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR