(2,6-Difluoropyridin-3-yl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

137483-1G 137483-500MG 137483-5G 137483-25G
77464-700EA 415.09 USD
77464-700 77477-480 77487-594 77498-378
(2,6-Difluoropyridin-3-yl)(phenyl)methanol ≥97%
(2,6-Difluoropyridin-3-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₉F₂NO
MW: 221.2027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34832856
CAS Number: 2921905-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR