1-(2,6-Difluoropyridin-3-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

137479-1G 137479-500MG 137479-5G 137479-25G
77464-696EA 415.09 USD
77464-696 77477-476 77487-590 77498-374
1-(2,6-Difluoropyridin-3-yl)cyclopentanol ≥97%
1-(2,6-Difluoropyridin-3-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₀H₁₁F₂NO
MW: 199.1972 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34832854
CAS Number: 2921714-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR