(2-Methoxyphenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

136560-1G 136560-500MG 136560-5G
77464-688EA 648.9 USD
77464-688 77477-468 77487-582
(2-Methoxyphenyl)(thiophen-2-yl)methanol ≥95%
(2-Methoxyphenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₂O₂S
MW: 220.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03211753
CAS Number: 356554-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR