2-(2,3-Dichlorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

136142-1G 136142-500MG 136142-5G 136142-25G
77464-684EA 279.13 USD
77464-684 77477-464 77487-578 77498-360
2-(2,3-Dichlorophenyl)propan-2-ol ≥95%
2-(2,3-Dichlorophenyl)propan-2-ol

Precaución: For research use only

Formula: C₉H₁₀Cl₂O
MW: 205.0811 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222270
CAS Number: 1340148-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR