4-Fluoro-3-(1-hydroxy-1-(thiophen-2-yl)ethyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

135329-1G 135329-500MG 135329-5G 135329-25G
77464-672EA 415.09 USD
77464-672 77477-452 77487-564 77498-346
4-Fluoro-3-(1-hydroxy-1-(thiophen-2-yl)ethyl)benzonitrile ≥97%
4-Fluoro-3-(1-hydroxy-1-(thiophen-2-yl)ethyl)benzonitrile

Precaución: For research use only

Formula: C₁₃H₁₀FNOS
MW: 247.288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34831613
CAS Number: 2969206-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR