1-(2,3-Difluorophenyl)-1-(thiophen-2-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

135301-1G 135301-500MG 135301-5G 135301-25G
77464-668EA 415.09 USD
77464-668 77477-448 77487-560 77498-342
1-(2,3-Difluorophenyl)-1-(thiophen-2-yl)ethanol ≥95%
1-(2,3-Difluorophenyl)-1-(thiophen-2-yl)ethanol

Precaución: For research use only

Formula: C₁₂H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD19626903
CAS Number: 1341571-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR