(2,6-Difluorophenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

135295-1G 135295-500MG 135295-5G 135295-25G
77464-664EA 397.58 USD
77464-664 77477-444 77487-556 77498-338
(2,6-Difluorophenyl)(thiophen-2-yl)methanol ≥95%
(2,6-Difluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈F₂OS
MW: 226.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787193
CAS Number: 1249018-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR