(5-Bromo-2-fluorophenyl)(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

135283-1G 135283-500MG 135283-5G 135283-25G
77464-660EA 394.49 USD
77464-660 77477-440 77487-552 77498-334
(5-Bromo-2-fluorophenyl)(thiophen-2-yl)methanol ≥97%
(5-Bromo-2-fluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈BrFOS
MW: 287.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787217
CAS Number: 1250357-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR