(2-Fluorophenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

135271-1G 135271-500MG 135271-5G 135271-25G
77464-656EA 169.95 USD
77464-656 77477-436 77487-548 77498-332
(2-Fluorophenyl)(thiophen-2-yl)methanol ≥95%
(2-Fluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₉FOS
Storage Temperature: Ambient
MDL Number: MFCD11097865
CAS Number: 860016-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR