1-(thiophen-2-yl)-1-(3-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

135257-1G 135257-500MG 135257-5G 135257-25G
77464-652EA 415.09 USD
77464-652 77477-432 77487-544 77498-328
1-(thiophen-2-yl)-1-(3-(trifluoromethyl)phenyl)ethanol ≥95%
1-(thiophen-2-yl)-1-(3-(trifluoromethyl)phenyl)ethanol

Precaución: For research use only

Formula: C₁₃H₁₁F₃OS
MW: 272.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19461676
CAS Number: 1282244-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR