(3-Chlorophenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

135251-1G 135251-500MG 135251-5G 135251-25G
77464-648EA 313.12 USD
77464-648 77477-428 77487-540 77498-324
(3-Chlorophenyl)(thiophen-2-yl)methanol ≥95%
(3-Chlorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₉ClOS
MW: 224.707 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03211667
CAS Number: 356552-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR