(4-Bromophenyl)(thiophen-2-yl)methanol ≥96%

Supplier: AOBChem USA

Warning

135243-1G 135243-500MG 135243-5G 135243-25G
77464-644EA 415.09 USD
77464-644 77477-424 77487-536 77498-320
(4-Bromophenyl)(thiophen-2-yl)methanol ≥96%
(4-Bromophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₉BrOS
MW: 269.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03211624
CAS Number: 40310-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR