2-Fluoro-4-methoxy-5-nitrobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

134691-1G 134691-500MG 134691-5G 134691-25G
77464-640EA 199.82 USD
77464-640 77477-420 77487-532 77498-318
2-Fluoro-4-methoxy-5-nitrobenzaldehyde ≥97%
2-Fluoro-4-methoxy-5-nitrobenzaldehyde

Precaución: For research use only

Formula: C₈H₆FNO₄
Storage Temperature: Ambient
MDL Number: MFCD27995599
CAS Number: 586412-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR