1-(3-(dimethylamino)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

134608-1G 134608-500MG 134608-5G
77464-636EA 497.49 USD
77464-636 77477-416 77487-528
1-(3-(dimethylamino)phenyl)-2-methylpropan-1-ol ≥95%
1-(3-(dimethylamino)phenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₉NO
MW: 193.2853 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34831127
CAS Number: 1512981-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR