2,4-Difluoronicotinic acid ≥95%

Supplier: AOBChem USA

Warning

13448-1G 13448-500MG 13448-5G 13448-25G
77464-632EA 245.14 USD
77464-632 77477-412 77487-524 77498-316
2,4-Difluoronicotinic acid ≥95%
2,4-Difluoronicotinic acid

Precaución: For research use only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031030
CAS Number: 849937-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR