(3-Ethoxy-5-formylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Ethoxy-5-formylphenylboronic acid

13391-1G 13391-5G 13391-25G
77464-628EA 67.98 USD
77464-628 77487-520 77498-312
(3-Ethoxy-5-formylphenyl)boronic acid ≥97%
(3-Ethoxy-5-formylphenyl)boronic acid

Precaución: For research use only

Formula: C₉H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD09038413
CAS Number: 1072952-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR