2-(o-tolyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

133317-1G 133317-500MG 133317-5G 133317-25G
77464-624EA 339.9 USD
77464-624 77477-408 77487-516 77498-308
2-(o-tolyl)propan-2-ol ≥97%
2-(o-tolyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD12067965
CAS Number: 7572-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR