4-Bromo-2-fluoro-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

13267-1G 13267-500MG 13267-5G 13267-25G
77464-620EA 284.28 USD
77464-620 77477-404 77487-512 77498-302
4-Bromo-2-fluoro-3-iodopyridine ≥97%
4-Bromo-2-fluoro-3-iodopyridine

Precaución: For research use only

Formula: C₅H₂BrFIN
Storage Temperature: Ambient
MDL Number: MFCD17014987
CAS Number: 917969-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR