2-Methyl-4-nitrophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

13242-1G 13242-500MG 13242-5G 13242-25G
77464-616EA 287.37 USD
77464-616 77477-402 77487-510 77498-298
2-Methyl-4-nitrophenylboronic acid pinacol ester ≥97%
2-Methyl-4-nitrophenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₃H₁₈BNO₄
Storage Temperature: Ambient
MDL Number: MFCD10698807
CAS Number: 883715-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR