4-Bromo-3-(methylsulfonyl)benzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

131766-1G 131766-5G 131766-25G
77464-612EA 60.77 USD
77464-612 77487-504 77498-294
4-Bromo-3-(methylsulfonyl)benzotrifluoride ≥95%
4-Bromo-3-(methylsulphonyl)benzotrifluoride

Precaución: For research use only

Formula: C₈H₆BrF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD03094286
CAS Number: 1215205-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR