tert-Butyl 4-hydroxy-4-(3-methoxyphenyl)piperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

130358-1G 130358-500MG 130358-5G 130358-25G
77464-604EA 152.44 USD
77464-604 77477-390 77487-492 77498-282
tert-Butyl 4-hydroxy-4-(3-methoxyphenyl)piperidine-1-carboxylate ≥95%
tert-Butyl 4-hydroxy-4-(3-methoxyphenyl)piperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₇H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD11849838
CAS Number: 871112-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR