6-Fluoro-1-methyl-1H-indole ≥97%

Supplier: AOBChem USA

Warning

130040-1G 130040-500MG 130040-5G 130040-25G
77464-600EA 338.87 USD
77464-600 77477-386 77487-488 77498-278
6-Fluoro-1-methyl-1H-indole ≥97%
6-Fluoro-1-methyl-1H-indole

Precaución: For research use only

Formula: C₉H₈FN
MW: 149.1649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09751642
CAS Number: 441715-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR